missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-1 Mouse, Clone: DSG1/1733, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1828-MSM3-P1
Additional Details : Weight : 0.01000kg
Specifications
Desmoglein-1 | |
Cell surface | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
DSG1 | |
Recombinant human desmoglein-1 protein fragment corresponding to intracellular domain | |
150kDa (precursor) ;160kDa (mature) | |
Protein A/G Purified | |
RUO | |
Primary | |
1828 |
Immunohistochemistry | |
DSG1/1733 | |
Unconjugated | |
NP_001933 | |
Mouse | |
IgG1 kappa | |
A431 or A-375 cells. Skin | |
100 μg | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
Monoclonal |