missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-1 Mouse, Clone: DSG1/1733, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £423.29
Specifications
| Antigen | Desmoglein-1 |
|---|---|
| Clone | DSG1/1733 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
Specifications
| Desmoglein-1 | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 1828 | |
| Recombinant human desmoglein-1 protein fragment corresponding to intracellular domain | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 150kDa (precursor) ;160kDa (mature) |
| DSG1/1733 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_001933 | |
| DSG1 | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cell surface | |
| A431 or A-375 cells. Skin |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title