missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmocollin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59227
This item is not returnable.
View return policy
Description
Desmocollin-3 Polyclonal specifically detects Desmocollin-3 in Human samples. It is validated for Western Blot.
Specifications
| Desmocollin-3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Cadherin family member 3, CDHF3HT-CP, desmocollin 3, desmocollin-4, DSC, DSC1, DSC2, DSC4desmocollin-3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1825 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q14574 | |
| DSC3 | |
| Synthetic peptides corresponding to DSC3(desmocollin 3) The peptide sequence was selected from the N terminal of DSC3. Peptide sequence MQENSLGPFPLFLQQVESDAAQNYTVFYSISGRGVDKEPLNLFYIERDTG. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Canine: 92%; Rat: 92%; Mouse: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction