missing translation for 'onlineSavingsMsg'
Learn More

(-)-Desethyl Methyl Etodolac, TRC

Product Code. 30364664
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364664 100 mg 100mg
30355568 25 mg 25mg
30348975 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364664 Supplier Toronto Research Chemicals Supplier No. TRCD289485100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109518-50-5
Molecular Formula C16H19NO3
Molecular Weight (g/mol) 273.33
Synonym USP Etodolac Related Compound A, (-)-8-Ethyl-1,3,4,9-tetrahydro-1-methylpyrano[3,4-b]indole-1-acetic Acid, Etodolac EP Impurity C
IUPAC Name 2-[(1R)-8-ethyl-1-methyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl]acetic acid
SMILES CCc1cccc2c3CCO[C@](C)(CC(=O)O)c3[nH]c12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H19NO3/c1-3-10-5-4-6-11-12-7-8-20-16(2,9-13(18)19)15(12)17-14(10)11/h4-6,17H,3,7-9H2,1-2H3,(H,18,19)/t16-/m1/s1
Formula Weight 273.1365
Chemical Name or Material (-)-Desethyl Methyl Etodolac
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.