CAS RN 109518-50-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(-)-Desethyl Methyl Etodolac, TRC
CAS: 109518-50-5 Molecular Formula: C16H19NO3 Molecular Weight (g/mol): 273.33 Synonym: USP Etodolac Related Compound A,(-)-8-Ethyl-1,3,4,9-tetrahydro-1-methylpyrano[3,4-b]indole-1-acetic Acid,Etodolac EP Impurity C IUPAC Name: 2-[(1R)-8-ethyl-1-methyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl]acetic acid SMILES: CCc1cccc2c3CCO[C@](C)(CC(=O)O)c3[nH]c12