missing translation for 'onlineSavingsMsg'
Learn More

Desethyl Felodipine, TRC

Product Code. 30357488
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357488 500 mg 500mg
30358676 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357488 Supplier Toronto Research Chemicals Supplier No. TRCD289240500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123853-39-4
Molecular Formula C16H15Cl2NO4
Molecular Weight (g/mol) 356.2
Synonym 1,4-Dihydro-2,6-dimethyl-4-(2',3'-dichlorophenyl)-5-carboxymethyl-3-pyridinecarboxylic acid, H 152/81, 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-Pyridinedicarboxylic Acid Monomethyl Ester
SMILES COC(=O)C1=C(C)NC(=C(C1c2cccc(Cl)c2Cl)C(=O)O)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H15Cl2NO4/c1-7-11(15(20)21)13(9-5-4-6-10(17)14(9)18)12(8(2)19-7)16(22)23-3/h4-6,13,19H,1-3H3,(H,20,21)
Formula Weight 355.0378
Chemical Name or Material Desethyl Felodipine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.