CAS RN 123853-39-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Desethyl Felodipine, TRC
CAS: 123853-39-4 Molecular Formula: C16H15Cl2NO4 Molecular Weight (g/mol): 356.2 Synonym: 1,4-Dihydro-2,6-dimethyl-4-(2',3'-dichlorophenyl)-5-carboxymethyl-3-pyridinecarboxylic acid,H 152/81,4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-Pyridinedicarboxylic Acid Monomethyl Ester SMILES: COC(=O)C1=C(C)NC(=C(C1c2cccc(Cl)c2Cl)C(=O)O)C