missing translation for 'onlineSavingsMsg'
Learn More

Deschloro-Zopiclone, TRC

Product Code. 30010988
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010988 10 mg 10mg
30007839 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010988 Supplier Toronto Research Chemicals Supplier No. TRCD28951510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1348046-61-6
Molecular Formula C17H18N6O3
Molecular Weight (g/mol) 354.36
Synonym 1-Piperazinecarboxylic acid, 4-methyl-, 6,7-dihydro-7-oxo-6-(2-pyridinyl)-5H-pyrrolo[3,4-b]pyrazin-5-yl ester, 6-Pyridin-2-yl-7-oxo-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-yl4-Methylpiperazine-1-carboxylate
SMILES CN1CCN(CC1)C(=O)OC2N(C(=O)c3nccnc23)c4ccccn4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H18N6O3/c1-21-8-10-22(11-9-21)17(25)26-16-14-13(19-6-7-20-14)15(24)23(16)12-4-2-3-5-18-12/h2-7,16H,8-11H2,1H3
Chemical Name or Material Deschloro-Zopiclone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.