CAS RN 1348046-61-6
CAS RN 1348046-61-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Deschloro-Zopiclone, TRC
CAS: 1348046-61-6 Molecular Formula: C17H18N6O3 Molecular Weight (g/mol): 354.36 Synonym: 1-Piperazinecarboxylic acid, 4-methyl-, 6,7-dihydro-7-oxo-6-(2-pyridinyl)-5H-pyrrolo[3,4-b]pyrazin-5-yl ester,6-Pyridin-2-yl-7-oxo-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-yl4-Methylpiperazine-1-carboxylate SMILES: CN1CCN(CC1)C(=O)OC2N(C(=O)c3nccnc23)c4ccccn4