missing translation for 'onlineSavingsMsg'
Learn More

Des(2-Hydroxypropyl) Gadoteridol, TRC

Product Code. 30357602
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357602 50 mg 50mg
30364715 10 mg 10mg
30360847 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357602 Supplier Toronto Research Chemicals Supplier No. TRCD29034050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112188-16-6
Molecular Formula C14H23GdN4O6
Molecular Weight (g/mol) 500.61
Synonym [1,4,7,10-tetraazacyclododecane-1,4,7-triacetato(3-)-N1,N4,N7,N10,O1,O4,O7]-Gadolinium, 1,4,7,10-Tetraazacyclododecane Gadolinium deriv., 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic Acid Gadolinium Complex, Gd-DO 3A
SMILES [O-]C(=O)CN12CC[NH]3CCN4(CC(=O)[O-])CCN(CC(=O)[O-])(CC1)[Gd+3]234

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H26N4O6.Gd/c19-12(20)9-16-3-1-15-2-4-17(10-13(21)22)6-8-18(7-5-16)11-14(23)24;/h15H,1-11H2,(H,19,20)(H,21,22)(H,23,24);/q;+3/p-3
Formula Weight 501.0859
Percent Purity >95%
Chemical Name or Material Des(2-Hydroxypropyl) Gadoteridol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.