CAS RN 112188-16-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Des(2-Hydroxypropyl) Gadoteridol, TRC
CAS: 112188-16-6 Molecular Formula: C14H23GdN4O6 Molecular Weight (g/mol): 500.61 Synonym: [1,4,7,10-tetraazacyclododecane-1,4,7-triacetato(3-)-N1,N4,N7,N10,O1,O4,O7]-Gadolinium,1,4,7,10-Tetraazacyclododecane Gadolinium deriv.,1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic Acid Gadolinium Complex,Gd-DO 3A SMILES: [O-]C(=O)CN12CC[NH]3CCN4(CC(=O)[O-])CCN(CC(=O)[O-])(CC1)[Gd+3]234