missing translation for 'onlineSavingsMsg'
Learn More

Des[2-[(2-Hydroxyethyl)amino]ethyl] Mitoxantrone (Mitoxantrone Impurity A), TRC

Product Code. 30136149
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136149 5 mg 5mg
30135100 50 mg 50mg
30010164 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136149 Supplier Toronto Research Chemicals Supplier No. TRCD2900055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89991-52-6
Molecular Formula C18 H19 N3 O5
Molecular Weight (g/mol) 357.36
Synonym 9,10-Anthracenedione, 1-amino-5,8-dihydroxy-4-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-, 1-Amino-5,8-dihydroxy-4-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione, 1-Amino-5,8-dihydroxy-4-[[2-[(2-hydroxyethyl)amino]ethyl]amino]anthracene-9,10-dione, 8-Amino-1,4-dihydroxy-5[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione, Mitoxantrone EP impurity A
IUPAC Name 1-amino-5,8-dihydroxy-4-[2-(2-hydroxyethylamino)ethylamino]anthracene-9,10-dione
SMILES Nc1ccc(NCCNCCO)c2C(=O)c3c(O)ccc(O)c3C(=O)c12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H19N3O5/c19-9-1-2-10(21-6-5-20-7-8-22)14-13(9)17(25)15-11(23)3-4-12(24)16(15)18(14)26/h1-4,20-24H,5-8,19H2
Formula Weight 357.1325
Chemical Name or Material 1-amino-5,8-dihydroxy-4-[[2-[(2-hydroxyethyl)amino]ethyl] amino]anthracene-9,10-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.