CAS RN 89991-52-6
CAS RN 89991-52-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Des[2-[(2-Hydroxyethyl)amino]ethyl] Mitoxantrone (Mitoxantrone Impurity A), TRC
CAS: 89991-52-6 Molecular Formula: C18 H19 N3 O5 Molecular Weight (g/mol): 357.36 Synonym: 9,10-Anthracenedione, 1-amino-5,8-dihydroxy-4-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-,1-Amino-5,8-dihydroxy-4-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione,1-Amino-5,8-dihydroxy-4-[[2-[(2-hydroxyethyl)amino]ethyl]amino]anthracene-9,10-dione,8-Amino-1,4-dihydroxy-5[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione,Mitoxantrone EP impurity A IUPAC Name: 1-amino-5,8-dihydroxy-4-[2-(2-hydroxyethylamino)ethylamino]anthracene-9,10-dione SMILES: Nc1ccc(NCCNCCO)c2C(=O)c3c(O)ccc(O)c3C(=O)c12