Learn More
Thermo Scientific™ Dermatophyte Medium with Phenol Red
Description
Selectively isolate dermatophyte fungi from the samples such as hair, skin scrapings and nails with Thermo Scientific™ Oxoid™ Dermatophyte Medium with Phenol Red. The near neutral pH of the medium is better for the growth of fungi. The often recommended acidic pH used to supress bacterial contaminants is replaced by addition of selective agents. Chloramphenicol and cycloheximide render the medium selective for dermatophytes inhibiting many saprophytic fungi, yeasts and bacterial skin flora1.
Use Dermatophyte Medium with Phenol Red for the isolation of dermatophyte fungi from samples such as hair, skin scrapings and nails.
- Inhibits the growth of contaminating saprophytic fungi, yeasts and bacterial skin flora1.
- Chloramphenicol and cycloheximide in the medium improve the isolation of dermatophytes, especially when the inoculum is heavily contaminated2, 3.
- Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Yellow / Orange |
| Content And Storage | 2°C to 10°C |
| Description | Dermatophyte Medium with Phenol Red |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 19.5g ± 1.0.g |
| Packaging Type | 10x90mm plates, film wrapped |
| Quantity | 10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.