Learn More
DEPDC6/Deptor Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | DEPDC6/Deptor |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18058765
|
Proteintech
CL488-20985-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18098195
|
Proteintech
CL488-20985-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DEPDC6/Deptor antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human DEPDC6/Deptor. This antibody recognizes human, mouse, rat antigen. The DEPDC6/Deptor antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| DEPDC6/Deptor | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| DEP domain containing 6, DEP.6, DEPDC6, Deptor, DKFZp564B1778, FLJ12341, FLJ12428, FLJ13854 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 64798 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.