Learn More
DEPDC6/Deptor Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-20985-100UL
Description
The DEPDC6/Deptor antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human DEPDC6/Deptor. This antibody recognizes human, mouse, rat antigen. The DEPDC6/Deptor antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| DEPDC6/Deptor | |
| 1000 μg/mL | |
| DEP domain containing 6, DEP.6, DEPDC6, Deptor, DKFZp564B1778, FLJ12341, FLJ12428, FLJ13854 | |
| 100 μL | |
| 64798 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |