missing translation for 'onlineSavingsMsg'
Learn More
Learn More
delta-Sarcoglycan Rabbit anti-Human, Mouse, Clone: 3P3R4, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £413.00
Specifications
| Antigen | delta-Sarcoglycan |
|---|---|
| Clone | 3P3R4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18314566
|
Novus Biologicals
NBP3-16776-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18317274
|
Novus Biologicals
NBP3-16776-100UL |
100 μg |
£413.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
delta-Sarcoglycan Monoclonal antibody specifically detects delta-Sarcoglycan in Human, Mouse samples. It is validated for Western BlotSpecifications
| delta-Sarcoglycan | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6444 | |
| IgG | |
| Affinity purified |
| 3P3R4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD | |
| A synthetic peptide corresponding to a sequence within amino acids 190-289 of human delta-Sarcoglycan (Q92629). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title