missing translation for 'onlineSavingsMsg'
Learn More
Learn More
delta-Sarcoglycan Rabbit anti-Human, Mouse, Clone: 3P3R4, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16776-100UL
This item is not returnable.
View return policy
Description
delta-Sarcoglycan Monoclonal antibody specifically detects delta-Sarcoglycan in Human, Mouse samples. It is validated for Western Blot
Specifications
| delta-Sarcoglycan | |
| Monoclonal | |
| Unconjugated | |
| 35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD | |
| A synthetic peptide corresponding to a sequence within amino acids 190-289 of human delta-Sarcoglycan (Q92629). | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 6444 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 3P3R4 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction