missing translation for 'onlineSavingsMsg'
Learn More
Learn More
delta Opioid R/OPRD1 Antibody (7G7), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | delta Opioid R/OPRD1 |
|---|---|
| Clone | 7G7 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029903
|
Novus Biologicals
NBP3-26447-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30025629
|
Novus Biologicals
NBP3-26447-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
delta Opioid R/OPRD1 Monoclonal antibody specifically detects delta Opioid R/OPRD1 in Human, Mouse samples. It is validated for Western Blot, ELISASpecifications
| delta Opioid R/OPRD1 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| delta opioid receptor 1, delta-type opioid receptor, D-OR-1, DOR-1, opioid receptor, delta 1, OPRD | |
| A synthesized peptide derived from Human delta Opioid R/OPRD1 [UniProt P41143] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7G7 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, GPCR, Membrane Trafficking and Chaperones | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 4985 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title