missing translation for 'onlineSavingsMsg'
Learn More
Learn More
delta Opioid R/OPRD1 Antibody (7G7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26447-50ul
This item is not returnable.
View return policy
Description
delta Opioid R/OPRD1 Monoclonal antibody specifically detects delta Opioid R/OPRD1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| delta Opioid R/OPRD1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 7G7 | |
| Western Blot 1:500-1:5000, ELISA | |
| delta opioid receptor 1, delta-type opioid receptor, D-OR-1, DOR-1, opioid receptor, delta 1, OPRD | |
| A synthesized peptide derived from Human delta Opioid R/OPRD1 [UniProt P41143] | |
| 50 μL | |
| Cancer, GPCR, Membrane Trafficking and Chaperones | |
| 4985 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction