missing translation for 'onlineSavingsMsg'
Learn More

Deiodo Amiodarone, TRC

Product Code. 30005576
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005576 10 mg 10mg
30010378 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005576 Supplier Toronto Research Chemicals Supplier No. TRCD22902510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85642-08-6
Molecular Formula C25 H30 I N O3
Molecular Weight (g/mol) 519.415
Synonym Methanone, (2-butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]-3-iodophenyl]-, (2-Butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone, L 6355, (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone, Deiodo Amiodarone, Desiodoamiodarone, (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone
IUPAC Name (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone
SMILES CCCCc1oc2ccccc2c1C(=O)c3ccc(OCCN(CC)CC)c(I)c3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C25H30INO3/c1-4-7-11-23-24(19-10-8-9-12-21(19)30-23)25(28)18-13-14-22(20(26)17-18)29-16-15-27(5-2)6-3/h8-10,12-14,17H,4-7,11,15-16H2,1-3H3
Formula Weight 519.127
Chemical Name or Material Deiodo Amiodarone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.