CAS RN 85642-08-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Deiodo Amiodarone, TRC
CAS: 85642-08-6 Molecular Formula: C25 H30 I N O3 Molecular Weight (g/mol): 519.415 Synonym: Methanone, (2-butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]-3-iodophenyl]-,(2-Butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone,L 6355,(2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone,Deiodo Amiodarone,Desiodoamiodarone,(2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone IUPAC Name: (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone SMILES: CCCCc1oc2ccccc2c1C(=O)c3ccc(OCCN(CC)CC)c(I)c3