missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Olmesartan, TRC

Product Code. 30357597
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357597 10 mg 10mg
30367549 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357597 Supplier Toronto Research Chemicals Supplier No. TRCD22996010MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172875-98-8
Molecular Formula C24 H24 N6 O2
Molecular Weight (g/mol) 428.49
Synonym 4-(1-Methylethenyl)-2-propyl-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid
IUPAC Name 5-prop-1-en-2-yl-2-propyl-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid
SMILES CCCc1nc(C(=C)C)c(C(=O)O)n1Cc2ccc(cc2)c3ccccc3c4nn[nH]n4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H24N6O2/c1-4-7-20-25-21(15(2)3)22(24(31)32)30(20)14-16-10-12-17(13-11-16)18-8-5-6-9-19(18)23-26-28-29-27-23/h5-6,8-13H,2,4,7,14H2,1,3H3,(H,31,32)(H,26,27,28,29)
Formula Weight 428.1961
Percent Purity >95%
Chemical Name or Material dehydro Olmesartan
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.