CAS RN 172875-98-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dehydro Olmesartan, TRC
CAS: 172875-98-8 Molecular Formula: C24 H24 N6 O2 Molecular Weight (g/mol): 428.49 Synonym: 4-(1-Methylethenyl)-2-propyl-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid IUPAC Name: 5-prop-1-en-2-yl-2-propyl-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid SMILES: CCCc1nc(C(=C)C)c(C(=O)O)n1Cc2ccc(cc2)c3ccccc3c4nn[nH]n4