missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Ivabradine, TRC

Product Code. 30134738
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134738 5 mg 5mg
17932525 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134738 Supplier Toronto Research Chemicals Supplier No. TRCD2291355MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1086026-31-4
Molecular Formula C27 H34 N2 O5
Molecular Weight (g/mol) 466.57
Synonym 2H-3-Benzazepin-2-one, 3-[3-[[[(7S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3-dihydro-7,8-dimethoxy-, 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3-dihydro-7,8-dimethoxy-2H-3-benzazepin-2-one, 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl](methyl)amino]propyl]-1,3-dihydro-7,8-dimethoxy-2H-3-benzazepin-2-one, Dehydro Ivabradine
IUPAC Name 3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methyl-methylamino]propyl]-7,8-dimethoxy-1H-3-benzazepin-2-one
SMILES COc1cc2CC(=O)N(CCCN(C)C[C@H]3Cc4cc(OC)c(OC)cc34)C=Cc2cc1OC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C27H34N2O5/c1-28(17-21-11-20-14-25(33-4)26(34-5)16-22(20)21)8-6-9-29-10-7-18-12-23(31-2)24(32-3)13-19(18)15-27(29)30/h7,10,12-14,16,21H,6,8-9,11,15,17H2,1-5H3/t21-/m1/s1
Formula Weight 466.2468
Chemical Name or Material Dehydro Ivabradine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.