missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEGS2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00 - £401.00
Specifications
| Antigen | DEGS2 |
|---|---|
| Dilution | Western Blot 1:400 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18330472
|
Novus Biologicals
NBP3-11918-25UG |
25 μg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392822
|
Bio-Techne
NBP3-11918-100UG |
100 μg |
£401.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DEGS2 Polyclonal antibody specifically detects DEGS2 in Human samples. It is validated for Western BlotSpecifications
| DEGS2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS | |
| 123099 | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:400 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C14orf66, chromosome 14 open reading frame 66, Degenerative spermatocyte homolog 2, degenerative spermatocyte homolog 2, lipid desaturase (Drosophila), DES2sphingolipid delta(4)-desaturase/C4-hydroxylase DES2, EC 1.14, FADS8, sphingolipid C4-hydroxylase/delta 4-desaturase, sphingolipid delta 4 desaturase/C-4 hydroxylase | |
| Synthetic peptide derived from human DEGS2 protein. (Uniprot: Q6QHC5) | |
| Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title