missing translation for 'onlineSavingsMsg'
Learn More

DEGS2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18392822 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392822 100 μg 100µg
18330472 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18392822 Supplier Bio-Techne Supplier No. NBP311918100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DEGS2 Polyclonal antibody specifically detects DEGS2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEGS2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:400
Formulation PBS
Gene Alias C14orf66, chromosome 14 open reading frame 66, Degenerative spermatocyte homolog 2, degenerative spermatocyte homolog 2, lipid desaturase (Drosophila), DES2sphingolipid delta(4)-desaturase/C4-hydroxylase DES2, EC 1.14, FADS8, sphingolipid C4-hydroxylase/delta 4-desaturase, sphingolipid delta 4 desaturase/C-4 hydroxylase
Host Species Rabbit
Immunogen Synthetic peptide derived from human DEGS2 protein. (Uniprot: Q6QHC5)
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 123099
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.