Learn More
PeproTech DEFB104A Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P268BT-500UG
Description
AA Sequence of recombinant protein: EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRTKP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Humn BD-4. Anti-Human BD-4-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hBD-4 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BD-4 (500-P268) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBD-4. Western Blot: To detect hBD-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBD-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P268BT-1MG will be provided as 2 x 500 μg
Defensins form a family of antimicrobial and cytotoxic peptides made by neutrophils. Defensins are short, processed peptide molecules that are classified by structure into three groups: alpha-defensins, beta-defensins and theta-defensins. All beta-defensin genes are densely clustered in four to five syntenic chromosomal regions. Chromosome 8p23 contains at least two copies of the duplicated beta-defensin cluster. This duplication results in two identical copies of defensin, beta 104, DEFB104A and DEFB104B, in head-to-head orientation. This gene, DEFB104A, represents the more centromeric copy.
Specifications
| DEFB104A | |
| Polyclonal | |
| Biotin | |
| DEFB104A | |
| BD-4; Beta-defensin 104; Beta-defensin 4; DEFB104; DEFB104A; DEFB104B; DEFB4; DEFB-4; defensin beta 104A; Defensin, beta 104; defensin, beta 104A; defensin, beta 4; hBD-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140596 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q8WTQ1 | |
| DEFB104A | |
| E.coli-derived Recombinant Human BD-4. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.