missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dectin-1/CLEC7A Rabbit anti-Mouse, Clone: 10, Novus Biologicals™
Rabbit Monoclonal Antibody
£307.00 - £458.00
Specifications
| Antigen | Dectin-1/CLEC7A |
|---|---|
| Clone | 10 |
| Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18281218
|
Novus Biologicals
NBP2-90491-50UL |
50 μg |
£307.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18265467
|
Novus Biologicals
NBP2-90491-100UL |
100 μg |
£458.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Dectin-1/CLEC7A | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
| Recombinant Mouse Dectin-1/CLEC7A protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 10 | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 64581 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title