missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dectin-1/CLEC7A Rabbit anti-Mouse, Clone: 10, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90491-100UL
This item is not returnable.
View return policy
Description
Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Dectin-1/CLEC7A | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| 10 | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
| Recombinant Mouse Dectin-1/CLEC7A protein | |
| 100 μg | |
| Signal Transduction | |
| 64581 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction