missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decorin, Mouse anti-Human, Clone: DCN/3523, Novus Biologicals™
Mouse Monoclonal Antibody
£226.00 - £405.00
Specifications
| Antigen | Decorin |
|---|---|
| Clone | DCN/3523 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18242458
|
Novus Biologicals
NBP2-79774-20UG |
20 μg |
£226.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18264116
|
Novus Biologicals
NBP2-79774-100UG |
100 μg |
£405.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Decorin Monoclonal specifically detects Decorin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Decorin | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B | |
| DCN | |
| IgG2b κ | |
| Protein A or G purified |
| DCN/3523 | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P07585, P07585 | |
| 1634 | |
| Recombinant human Decorin protein fragment (aa212-336) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title