missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decorin, Mouse anti-Human, Clone: DCN/3523, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79774-20UG
This item is not returnable.
View return policy
Description
Decorin Monoclonal specifically detects Decorin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Decorin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P07585, P07585 | |
| DCN | |
| Recombinant human Decorin protein fragment (aa212-336) (exact sequence is proprietary) | |
| 20 μg | |
| Cancer | |
| 1634 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| DCN/3523 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction