missing translation for 'onlineSavingsMsg'
Learn More

Cytiva DEAE-Sepharose™ Media

Product Code. 11584935 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11584935 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11584935 Supplier Cytiva Supplier No. 17071001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Developed for process-scale chromatography

  • Weak anion exchanger with excellent flow properties
  • High capacity for proteins of all pI values
  • Ion exchange group is diethylaminoethyl, which remains charged and maintains consistently high capacities over the entire working range, pH 3 to 9

TRUSTED_SUSTAINABILITY

Specifications

Buffer 20mM N-methylpiperazine, 20mM Piperazine, 20mM L-Histidine, 20mM Bis-Tris, 20mM Bis-Tris Propane, 20mM Triethanolamine, 20mM Tris, 50mM N-methyldiethanolamine, 20mM Diethanolamine at pH 8.4, 20mM 1,3-Diaminopropane, 20mM Ethanolamine, 20mM Piperazine, 20mM 1,3-Diaminopropionate
Product Type DEAE Sepharose CL-6B media
Quantity 500 mL
Flow Rate <150cm/hr.
Ligand Type Diethylaminoethyl
pH Range 3 to 9
Particle Size 45 to 165μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
For Use With (Application) Process scale chromato graphy
Matrix DEAE-Sepharose
Show More Show Less
compliance-icons
Product Identifier
  • DEAE Sepharose CL-6B
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.