missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX3 Mouse anti-Human, Unconjugated, Clone: 1D3A1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DDX3 |
|---|---|
| Clone | 1D3A1 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17442413
|
Proteintech
67915-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17432413
|
Proteintech
67915-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DDX3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDX3. This antibody recognizes human, rat antigen. The DDX3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| DDX3 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human, Rat | |
| IgG2b |
| 1D3A1 | |
| Monoclonal | |
| Mouse | |
| 1654 | |
| Primary |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit