Learn More
DDX3 Mouse anti-Human, Unconjugated, Clone: 1D3A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67915-1-IG-20UL
This item is not returnable.
View return policy
Description
The DDX3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDX3. This antibody recognizes human, rat antigen. The DDX3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| DDX3 | |
| Monoclonal | |
| Unconjugated | |
| 20 μL | |
| 1654 | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 1D3A1 | |
| Mouse | |
| Primary | |
| Human, Rat |