missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDB2 Recombinant Rabbit Monoclonal Antibody (23GB2805), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | DDB2 |
|---|---|
| Clone | 23GB2805 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089974
|
Invitrogen™
MA553280 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089659
|
Invitrogen™
MA553281 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDB2 Recombinant Monoclonal antibody specifically detects DDB2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| DDB2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92466 | |
| 1643 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2805 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610043A19Rik; damage specific DNA binding protein 2; damage-specific DNA binding protein 2, 48kDa; damage-specific DNA-binding protein 2; DDB p48 subunit; DDB2; DDBb; DNA damage-binding protein 2; UV-damaged DNA-binding protein 2; UV-DDB 2; UV-DDB2; xeroderma pigmentosum group E protein; XPE | |
| A synthesized peptide derived from human DDB2 (350-427AA). | |
| Primary | |
| -20°C | |
| DDB2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title