missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDB2 Recombinant Rabbit Monoclonal Antibody (23GB2805), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553281
This item is not returnable.
View return policy
Description
Damaged DNA binding protein (DDB) is a heterodimer composed of two subunits, p127 and p48, which are designated DDB1 and DDB2, respectively. The DDB heterodimer is involved in repairing DNA damaged by ultraviolet light. Specifically, DDB, also designated UV-damaged DNA binding protein (UV-DDB), xeroderma pigmentosum group E binding factor (XPE-BF) and hepatitis B virus X-associated protein 1 (XAP-1), binds to damaged cyclobutane pyrimidine dimers (CPDs). Mutations in the DDB2 gene are implicated as causes of xeroderma pigmentosum group E, an autosomal recessive disease in which patients are defective in nucleotide excision DNA repair. XPE is characterized by hypersensitivity of the skin to sunlight with a high frequency of skin cancer as well as neurologic abnormalities. The hepatitis B virus (HBV) X protein interacts with DDB1, which may mediate HBx transactivation.
Specifications
| DDB2 | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92466 | |
| DDB2 | |
| A synthesized peptide derived from human DDB2 (350-427AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2805 | |
| Unconjugated | |
| DDB2 | |
| 2610043A19Rik; damage specific DNA binding protein 2; damage-specific DNA binding protein 2, 48kDa; damage-specific DNA-binding protein 2; DDB p48 subunit; DDB2; DDBb; DNA damage-binding protein 2; UV-damaged DNA-binding protein 2; UV-DDB 2; UV-DDB2; xeroderma pigmentosum group E protein; XPE | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1643 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction