missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDAH2 Recombinant Rabbit Monoclonal Antibody (23GB3820), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | DDAH2 |
|---|---|
| Clone | 23GB3820 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087525
![]() |
Invitrogen™
MA552572 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088851
|
Invitrogen™
MA552573 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDAH2 Recombinant Monoclonal antibody specifically detects DDAH2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| DDAH2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O95865, Q6MG60, Q99LD8 | |
| 23564, 294239, 51793 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3820 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110003M04Rik; AU019324; AW413173; Clone 7u; DADB-110M10.5; Ddah; DDAH2; DDAH-2; DDAHII; dimethylargininase-2; dimethylarginine dimethylaminohydrolase 2; dimethylarginine dimethylaminohydrolase II; epididymis secretory protein Li 277; G6a; HEL-S-277; N(G),N(G)-dimethylarginine dimethylaminohydrolase 2; NG30; OTTHUMP00000029307; OTTHUMP00000062666; OTTHUMP00000174406; Protein G6a; S-phase protein; testis tissue sperm-binding protein Li 54e | |
| A synthetic peptide of human DDAH2 (1-75AA). | |
| Primary | |
| -20°C | |
| Ddah2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title