missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDAH2 Recombinant Rabbit Monoclonal Antibody (23GB3820), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552573
This item is not returnable.
View return policy
Description
This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.
Specifications
| DDAH2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Ddah2 | |
| 1110003M04Rik; AU019324; AW413173; Clone 7u; DADB-110M10.5; Ddah; DDAH2; DDAH-2; DDAHII; dimethylargininase-2; dimethylarginine dimethylaminohydrolase 2; dimethylarginine dimethylaminohydrolase II; epididymis secretory protein Li 277; G6a; HEL-S-277; N(G),N(G)-dimethylarginine dimethylaminohydrolase 2; NG30; OTTHUMP00000029307; OTTHUMP00000062666; OTTHUMP00000174406; Protein G6a; S-phase protein; testis tissue sperm-binding protein Li 54e | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23564, 294239, 51793 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3820 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95865, Q6MG60, Q99LD8 | |
| Ddah2 | |
| A synthetic peptide of human DDAH2 (1-75AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction