missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCAF4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52862
This item is not returnable.
View return policy
Description
DCAF4 Polyclonal specifically detects DCAF4 in Human samples. It is validated for Western Blot.
Specifications
| DCAF4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DDB1 and CUL4 associated factor 4, DDB1- and CUL4-associated factor 4, DKFZp434K114, MGC20547, MGC46524, WD repeat domain 21, WD repeat domain 21A, WD repeat-containing protein 21A, WDR21, WDR21A | |
| Rabbit | |
| 43 kDa | |
| 100 μL | |
| Primary | |
| Rat: 86%; Bovine: 86%; Guinea pig: 86%; Mouse: 79%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8IV10 | |
| DCAF4 | |
| Synthetic peptides corresponding to WDR21A(WD repeat domain 21A) The peptide sequence was selected from the middle region of WDR21A. Peptide sequence GHRQSFGTNSDVLAQQFALMAPLLFNGCRSGEIFAIDLRCGNQGKGWKAT. | |
| Affinity purified | |
| RUO | |
| 26094 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction