Learn More
Masterflex™ DB15 Male Connector
Click to view available options
For Use With:
Masterflex™ Drives
Unit Size:
Each
Description
- Combine connectors with 20-22 gauge shielded wire to create your own interface cable
- Maximum cable length is 300 m
Specifications
Specifications
| For Use With (Application) | Creating Your Own Cable |
| For Use With | Masterflex™ Drives |
| Inlet Connection (English) | DB15 |
| Type | Male Connector |