Learn More
Emma Safety Footwear Daytona Safety Shoes
Ideal for warm, enclosed environments. Features Emma's unique Hydro-Tec™ Sanitized Silver technology, removing moisture and perspiration instantly. Synthetic, reinforced toes and super swift Velcro fasteners make the Daytona breathable, wearable and incredibly comfortable.
Brand:  Emma Safety Footwear MM78654045
Additional Details : Weight : 1.60600kg
 View more versions of this product
Product Code. 15959041
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • S1-Safety shoe for dry working conditions
  • Nubuck construction
  • GripForce™ Easy Twist Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Velcro closure
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Black, Gray
Hydro-Tec™ Sanitized Silver
45
Velcro
Daytona
Nubuck
Steel