missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DAU 5884 hydrochloride
GREENER_CHOICE

Product Code. 30001314
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001314 10 mg 10mg
18014742 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001314 Supplier Tocris Bioscience™ Supplier No. 2096/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

M3 receptor antagonist

Selective muscarinic M3 receptor antagonist. In bovine tracheal smooth muscle, inhibits methacholine-dependent effects on cell proliferation and muscle contractility.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131780-48-8
Molecular Formula C17H22ClN3O3

Specifications

Quantity 10 mg
Formula Weight 351.83
Purity >99%
Molecular Weight (g/mol) 351.831
Chemical Name or Material DAU 5884 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.