accessibility menu, dialog, popup

UserName

CAS RN 131780-48-8

CH3NOONNHOHCl

CAS RN 131780-48-8

IUPAC Name: 8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-oxo-1,2,3,4-tetrahydroquinazoline-3-carboxylate hydrochloride
Synonyms: 8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-oxo-1,4-dihydroquinazoline-3-carboxylate hydrochloride
Molecular Weight (g/mol): 351.83
Molecular Formula: C17H22ClN3O3
InChi Key: FDERDDSQHZRNGC-UHFFFAOYNA-N
SMILES: Cl.CN1C2CCC1CC(C2)OC(=O)N1CC2=CC=CC=C2NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results