missing translation for 'onlineSavingsMsg'
Learn More

DARC Antibody (18B11), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen DARC
Clone 18B11
Dilution Flow Cytometry 1:50-1:200, ELISA
Applications Flow Cytometry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043320
View Documents
Novus Biologicals
NBP3-26634-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043609
View Documents
Novus Biologicals
NBP3-26634-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DARC Monoclonal antibody specifically detects DARC in Human samples. It is validated for Flow Cytometry, ELISA
Specifications

Specifications

DARC
Flow Cytometry 1:50-1:200, ELISA
Monoclonal
Purified
RUO
Human
CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor
A synthesized peptide derived from Human DARC [UniProt Q16570]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
18B11
Flow Cytometry, ELISA
Unconjugated
Rabbit
GPCR, Immunology, Innate Immunity
PBS, pH 7.4, 150mM NaCl and 50% glycerol
2532
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.