missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DARC Antibody (18B11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26634-100ul
This item is not returnable.
View return policy
Description
DARC Monoclonal antibody specifically detects DARC in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| DARC | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 18B11 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor | |
| A synthesized peptide derived from Human DARC [UniProt Q16570] | |
| 100 μL | |
| GPCR, Immunology, Innate Immunity | |
| 2532 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction