missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DAPT

Description
DAPT is a γ-secretase inhibitor. DAPT reduces Aβ40 and Aβ42 levels in human primary neuronal cultures (IC50 values are 115 and 200 nM for total Aβ and Aβ42 respectively) and in brain extract, cerebrospinal fluid and plasma in vivo. DAPT has no effect on APPα and APPβ levels. DAPT blocks Notch signaling in hybrid human-mouse fetal thymus organ culture (FTOC) and causes ESCs to commit to neuronal differentiation. DAPT can be used in a small molecule cocktail to derive cortical neurons from hPSCs and to maintain hepatocytes in culture. DAPT also promotes the formation of cone photoreceptors in retinal organoids.
For more information about how DAPT may be used, see our protocols: Accelerated Induction of Cortical Neurons from hiPSCs, Human iPSC-derived Myoblasts, Generating Midbrain Dopaminergic Neurons from hPSCs .
DAPT synthesized to Ancillary Material Grade also available.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 432.46 |
| Purity | >99% |
| Molecular Weight (g/mol) | 432.468 |
| Chemical Name or Material | DAPT |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?