missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Dapoxyl™ Carboxylic Acid, Succinimidyl Ester

Product Code. 11510186
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11510186 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11510186 Supplier Molecular Probes™ Supplier No. D10161

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Dapoxyl™ carboxylic acid, succinimidyl ester

The amine-reactive Dapoxyl™ succinimidyl ester can be conjugated to amine-containing biomolecules of interest to create an environmentally sensitive probe. The fluorescence of Dapoxyl™ derivatives is very environment sensitive and their fluorescence in water is very low, suggesting that Dapoxyl™conjugates may be useful as probes for conformational changes, denaturation, phosphorylation state and phase changes. Dapoxyl™ dye exhibits a huge Stokes shifts (up to ∼200 nm) and large extinction coefficient in some solvents.
TRUSTED_SUSTAINABILITY

Specifications

Description Dapoxyl™ Carboxylic Acid, Succinimidyl Ester, 5mg, Orange, Solid, C22H19N3O5 Molecular Formula, 405.4092 Molecular Weight, Dapoxyl™ Label, Amine Chemical Reactivity, 395/600nm Excitation/Emission Wavelength, Used for Labeling Chemistry, Protei
Formulation Solid
For Use With (Application) Labeling chemistry, protein labeling and crosslinking
Quantity 5 mg
Product Type Dapoxyl™ Carboxylic Acid, Succinimidyl Ester
Molecular Weight (g/mol) 405.41kDa
Content And Storage -5°C to -30°C
Label or Dye Dapoxyl™
Color Orange
Emission 600
Excitation 395
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.