missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAPK3/ZIPK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | DAPK3/ZIPK |
|---|---|
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
DAPK3/ZIPK Polyclonal specifically detects DAPK3/ZIPK in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.Specifications
| DAPK3/ZIPK | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS with 0.02% Sodium Azide | |
| DAP kinase 3, DAP-like kinase, death-associated protein kinase 3, dlk, EC 2.7.11, EC 2.7.11.1, ZIP, ZIP kinase isoform, ZIPKFLJ36473, ZIP-kinase | |
| DAPK3 | |
| IgG | |
| Affinity Purified | |
| 27 kDa |
| 1 mg/ml | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| BAA81746 | |
| 1613 | |
| Antibody was raised against a peptide corresponding to amino acids near the center of human ZIP kinase. The immunogen is located within amino acids 270 - 320 of ZIPK. Amino Acid Squence: RRRNVRGEDSGRKPERRRLK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title