missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAPK3/ZIPK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76962-0.025mg
This item is not returnable.
View return policy
Description
DAPK3/ZIPK Polyclonal specifically detects DAPK3/ZIPK in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.
Specifications
| DAPK3/ZIPK | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DAP kinase 3, DAP-like kinase, death-associated protein kinase 3, dlk, EC 2.7.11, EC 2.7.11.1, ZIP, ZIP kinase isoform, ZIPKFLJ36473, ZIP-kinase | |
| Rabbit | |
| 27 kDa | |
| 0.025 mg | |
| Apoptosis | |
| 1613 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL | |
| BAA81746 | |
| DAPK3 | |
| Antibody was raised against a peptide corresponding to amino acids near the center of human ZIP kinase. The immunogen is located within amino acids 270 - 320 of ZIPK. Amino Acid Squence: RRRNVRGEDSGRKPERRRLK | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction